Revco™ RDE Series –40 °C Ultra-Low Temperature Freezers with Racks and Boxes, Thermo Scientific
Supplier: Thermo Scientific
Revco®
RDE50040FARK RDE40040LARK RDE50040LDRK RDE60040FDRK RDE40040FARK RDE40040LDRK RDE50040FDRK RDE50040FARK RDE50040LARK RDE50040LDRK RDE60040LARK RDE40040LARK RDE50040LARK RDE60040FARK RDE40040FDRK RDE60040LARK RDE60040FDRK RDE40040LDRK RDE50040FDRK RDE40040FDRK RDE60040LDRK RDE60040FARK RDE60040LDRK RDE40040FARK
76415-568EA 16877.86 USD
CA76415-536 76415-568 CA76415-588 CA76415-560 CA76415-524 76415-574 76415-544 76415-536 76415-580 76415-588 CA76415-596 CA76415-568 CA76415-580 76415-552 CA76415-530 76415-596 76415-560 CA76415-574 CA76415-544 76415-530 76415-604 CA76415-552 CA76415-604 76415-524
Revco™ RDE Series –40 °C Ultra-Low Temperature Freezers with Racks and Boxes, Thermo Scientific
Freezers Ultra Low Temperature Freezers
Dependable –40 °C storage, powered by H-drive for operational savings and sustainability.
  • Low energy
  • Low peak variation
  • Fast recovery after door opening
  • Long warm up time
  • User friendly and quiet
Designed to keep your samples safe. Limit environmental impact and lower HVAC costs.
Certificaciones: SNAP Compliant, cULus certified.
Información de suministro: Package include one ULT freezer and two shelves of sliding drawer racks and boxes.