VACUUBRAND® Replacement Tubing for VHCpro™ Fluid Aspiration Hand Controller, BrandTech® Scientific
Supplier: BrandTech
VACUUBRAND®
20636156
89239-628EA 39.61 USD
89239-628
VACUUBRAND® Replacement Tubing for VHCpro™ Fluid Aspiration Hand Controller, BrandTech® Scientific
Tubing Silicone Tubing
Replace the worn or sticky tubing from your VACUUBRAND® BVC basic, control or professional fluid aspiration system.
  • Sold by the meter
Made from high quality silicone, this OEM replacement tubing will bring new life to your VHCpro hand controller.
Información para pedidos: Minimum order length 2 meters.