Dura-Mag™ Stirrers, Magnetic, 2-Position, 230 V
Supplier: Chemglass
CLS-4100-A2E
76333-478EA 5613.1 USD
76333-478
Dura-Mag™ Stirrers, Magnetic, 2-Position, 230 V
Stirrers Magnetic Stirrers, Multi-position
Digital, front-end, magnetic stirrers are designed for both cell culture in spinner flasks as well as general laboratory procedures requiring the use of magnetic stirring equipment.
  • 230 V model
Dura-Mag™ stirrers feature brushless, maintenance-free DC motors with drive assemblies designed for quiet operation. Stainless steel covers are stamped to show the vessel positions for accurate placement and optimum performance. Dura-Mag™ 2-position, low profile chassis measure 9.5W×22.5D×3.5"H. Designed to fit side-by-side in most incubators. Maximum flask size is (2) 8 L vessels. Optional factory installed motion alarm available. Speed range: 25 to 400 rpm.
Precaución: 230 V model.