Corepoint Scientific™ Temperature Monitoring Devices, Horizon Scientific
Supplier: Horizon Scientific
NS-TMD-18 NS-DDL1-18 NS-WF-DDL-18 NS-DDL2-18
76402-948EA 269 USD
76402-948 76402-952 76402-954 76402-950
Corepoint Scientific™ Temperature Monitoring Devices, Horizon Scientific
Protect your valuable samples by using only calibrated temperature monitoring devices.
  • Complies with current CDC, VFC, and CFR guidelines
  • Three years certification of calibration
  • High/low temperature, visual and audible alarms
  • Buffered probe in sample simulated solution
  • °F/°C switchable readout
CDC recommends (and VFC requires) using only calibrated temperature monitoring devices with a certificate of calibration provides a range of temperature monitoring solutions that are compliant with CDC, VFC and state boards of pharmacy guidelines.