NUCLEOSIL® SA, HPLC Columns, Hichrom Limited
Supplier: Hichrom Limited
NC100-5SA-250A NC10010SA100A NC100-5SA-150A NC10010SA150A NC1005SA150AM NC1005SA250AS NC1005SA125AF NC100-5SA-50AF NC100-5SA-125A NC10010SA10C5 NC100-5SA-100A NC1005SA100AF NC100-10SA-50A NC1005SA125AS NC1005SA10C5SK NC1005SA10CM5 NC100-5SA-10C5 NC10010SA100AF NC1005SA100AS NC100-5SA-200A NC10010SA300A NC1005SA150AS NC100-5SA-50A NC1005SA250AF NC100-5SA-50AM NC100-10SA250A NC1005SA200AF NC100-5SA-50AS NC1005SA150AF NC10010SA250AF NC1005SA100AM
76401-308EA 571.84 USD
76401-308 76400-940 76401-306 76400-788 76400-942 76401-304 76401-294 76401-096 76401-094 76401-092 76401-090 76401-314 76401-312 76400-934 76401-310 76400-936 76401-298 76400-938 76401-296 76401-318 76400-950 76401-316 76400-952 76400-790 76401-086 76400-944 76401-302 76400-946 76401-300 76400-948 76401-088
NUCLEOSIL® SA, HPLC Columns, Hichrom Limited
These are packed with classical spherical porous silica available in a wide range of pore sizes.
  • Sulphonic acid (SA) functional group
  • Good mechanical stability
  • Good batch to batch reproducibility
The 100 and 120 Å have similar relative retention characteristics, however the 100 Å has the greater capacity whilst the 120 Å exhibits a lower backpressure.