NUCLEOSIL® C8, HPLC Columns, Hichrom Limited
Supplier: Hichrom Limited
NC100-5C8-10C5 NC1007C8300AF NC100-5C8-100A NC100-10C8-50A NC100-7C8-10C5 NC1005C810C5SK NC1005C8300AF NC100-7C8-100A NC1005C8300AS NC10010C8100A NC100-7C8-250A NC100-5C8-250A NC1005C8125AF NC1007C8250AF NC1005C8150AS NC100-5C8-50AS NC100-7C8-50A NC100-5C8-50AF NC1005C8125AS NC1005C8150AF NC100-5C8-125A NC100-7C8-150A NC1005C8250AF NC10010C8250AF NC100-5C8-150A NC1005C8100AS NC10010C8150A NC100-7C8-125A NC100-5C8-50A NC100-5C8-200A NC1005C8250AS NC100-7C8-300A NC10010C810C5 NC10010C8200A NC1005C8100AF NC10010C8250A NC1007C8125AF NC10010C8125AF
76400-784EA 539.6 USD
76400-784 76400-862 76401-228 76400-864 76401-052 76401-050 76401-358 76400-878 76401-236 76400-998 76401-356 76401-234 76401-058 76401-354 76401-056 76401-232 76401-230 76401-054 76400-870 76400-872 76400-874 76400-994 76401-218 76400-876 76400-996 76401-216 76401-062 76401-060 76401-006 76401-226 76400-866 76401-224 76401-004 76400-868 76401-002 76401-222 76401-220 76401-000
NUCLEOSIL® C8, HPLC Columns, Hichrom Limited
Packed with classical spherical porous silica available in a wide range of pore sizes.
  • Good mechanical stability
  • Good batch to batch reproducibility
The 100 and 120 Å have similar relative retention characteristics, however the 100 Å has the greater capacity whilst the 120 Å exhibits a lower backpressure.