Pneumatic Adjustable-Height Standing Desk, 48", Luxor

Supplier: Luxor
SPN48F-BK/TK
76203-756EA 1284.36 USD
76203-756
Pneumatic Adjustable-Height Standing Desk, 48", Luxor
Furniture Office Furniture
Quickly deliver collaborative and productive comfort to your workday by seamlessly alternating between sitting and standing with the Luxor pneumatic adjustable height standing desk.

  • Lever-activated pneumatic air cylinder easily adjusts height anywhere between 27.5" and 44.5"
  • Raises and lowers up to 150 lbs., easily housing a CPU, monitors and other office accessories
  • Sturdy, scratch-resistant desktop with a teak finish
  • Constructed with two sturdy steel legs, each reinforced with durable inner legs
  • Chamfered edges keep forearms comfortable while typing and working

Its lever-activated pneumatic air cylinder lets you easily alter the desk's height between 27.5" and 44.5", smoothly lifting and lowering your monitors, computer and other office accessories. With two sturdy steel legs, reinforced inner legs and a high-quality laminate finish, this stand-up desk is built to last. With no power outlets required for use, it's perfect for any corporate or home office space.

Certificaciones: Meets ANSI/BIFMA standards.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR