GAMMEX® Cut-Resistant Glove Liners, Ansell
Supplier: Ansell
Gammex®
5789912 5789911 5789914 5789913
75849-350CS 960.15 USD
75849-350 75853-846 75856-924 75856-922
GAMMEX® Cut-Resistant Glove Liners, Ansell
Gloves
Non-latex, Dyneema® yarn cut-resistant glove liners provide adequate cut resistance and protection during rigorous procedures.
  • Moderate level of cut resistance protection
  • Reduces the risk of cut-related injuries when handling sharp instruments
  • Sterile and single use-regulated by the FDA as Class 1 medical device
GAMMEX® cut-resistance glove liners are, a strong, high-denisty fiber that provides adequate cut resistance protection during rigorous procedures while optimizing dexterity, comfort, and fit. Glove liners are classified as personal protection equiment (PPE) and not as medical devices. These non-latex, single use cut-resistant liners are sterile and chemical accelerator-free. The non-latex and chemical accelerator-free design helps prevent latex Type 1 allergies and minimizes chemical Type IV allergies and sensitivities. They are recommended to be worn between two powder-free gloves when used for surgical procedures.
Certificaciones: Product Standards: EN 388; EN 420; EN ISO 13997; ASTM F1790. Quality/Environmental Standards: ISO 9001; ISO 13485; ISO 14001; ISO 11137-Part 1; EN 556. ANZI/ISEA 105 cut level 2; EN ISO 13997 cut resistance level B; EN388 abrasion level 3.
Envase: Gloves are packaged five pairs per box.
Precaución: Warning: No glove liner provides complete protection against cuts. Use with care when handling sharp instruments. The glove liner offers protection against laceration-type and scalpel injuries, but not needle stick injuries.