Clarity Drugs of Abuse Urine Single Dip Test, Clarity Diagnostics
Supplier: Clarity Diagnostics
Clarity
DTG-BARC DTG-COC300C DTG-AMP1000C DTG-MTDC DTG-UALC DTG-MAMP1000C DTG-MDMAC DTG-THCC DTG-BUPC DTG-OPI300C DTG-OXYC DTG-PCPC DTG-OPI2000C DTG-COC150C DTG-BZD300C
95064-107CS 1474.13 USD
95064-107 95064-109 95064-105 95064-099 95064-101 95064-079 95064-089 95064-111 95064-095 89168-282 95064-083 95064-097 95064-085 95064-091 95064-081
Clarity Drugs of Abuse Urine Single Dip Test, Clarity Diagnostics
Drug Tests
Single dip panel test for drugs of abuse detection in urine samples.
  • Results in 5 minutes
  • Results can be photocopied
  • 25 tests/box
  • 16-18 months dating
Guaranteed to be accurate and built to prevent errors such as urine contamination and errors in results.
Precaución: Forensic use only.