Extech™ SMD Component Fixture for LCR Meter, Flir®
Supplier: Flir
LCR205
89370-150EA 274.35 USD
89370-150
Extech™ SMD Component Fixture for LCR Meter, Flir®
Electrical Meters Digital Multimeters
The component is useful for testing tasks like impedance evaluations on SMDs.
  • Adjustment range 0 to 0.375" (0.95 cm)
  • Terminals: 3 blades (high, low, guard)
  • Adjustment knob
Optional chip component fixture for use with 89370-146 LCR meter. The unit features three terminals for High, Low and Guard as well as an adjustment knob to secure measurement contacts to components being tested.
Certificaciones: CE Marked.