Wall Cavity Probe, Flir®
Supplier: Flir
MR06
10818-360EA 374.13 USD
10818-360
Wall Cavity Probe, Flir®
Electrodes Other Electrodes
Designed for use with the FLIR MR series moisture meters. Comes with two sets of pins.
  • Insulated pins designed for increased durability
  • Rubberized ergonomic grips reduce slippage
  • Detachable cord with a durable BNC connection
  • Pins included: 1 pair of 4" pins, 1 pair of 6" pins
  • Impact plate to drive pins through a drywall
The MR06 910818-360) is a Wall Cavity Probe. Designed for use with the FLIR MR series moisture meters. Comes with two sets of pins.