Midstream Collection Kits for Urinalysis, Medegen Medical Products
Supplier: Medegen Medical Products
4150A 4152A 4143A 4141A M4625
76329-618CS 135.73 USD
76329-618 76329-616 76329-610 76329-614 76329-612
Midstream Collection Kits for Urinalysis, Medegen Medical Products
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Obtain clean catch urine specimens with ease using convenient collection kits with either tab rings or anatomically contoured funnels.
  • Each 4 oz. specimen container is calibrated in 10 ml and 0.25 oz. increments up to 120 ml
  • Labels have lines for names, room number, doctor, date, time and type of specimen
  • UriAid™ eliminates need for transfer of urine specimen and can be used in pneumatic tube systems
All pertinent supplies for clean catch specimens are placed within the package. All midstream catch kits and UriAid™ come with easy-to-read instructions. UriAid™ is a single use funnel that screws onto the sterile, leak resistant specimen container that comes with this kit.