Hybridization Solution, QuikHyb, Agilent Technologies
Supplier: Agilent Technologies
201221 201221
99900-676EA 1054.78 USD
99900-676 CA99900-676
Hybridization Solution, QuikHyb, Agilent Technologies
Buffers Electrophoresis Buffers Hybridization Buffers
QuikHyb hybridization solution is ideal for use in Southern, Northern, slot-blot and plaque-lift formats using randomly labeled radioactive and nonradioactive nucleic acid probes.
This easy-to-use solution reduces the time required for hybridization from the customary 12 to 24 hours down to 1 to 2 hours, resulting in sensitive detection of target sequences.
Precaución: For in vitro use only.