Sodium hydroxide 97.0-100.5% alkali metal, pellets, GenAR® NF, FCC, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIALS US
Danger

Synonyms: Caustic soda

Management of Change (MOC) category = R

BS20-D3 MACRBS20-SA
JTBS20-10EA 227.08 USD
JTBS20-10 JTBS20-D1 JTBS20-D3 JTBS20-ST MKBS20SA
Sodium hydroxide 97.0-100.5% alkali metal, pellets, GenAR® NF, FCC, Macron Fine Chemicals™
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II

Order Now

Specification Test Results

Meets F.C.C. Requirements
Meets J.P. Chemical Specifications
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets N.F. Requirements
Food GMP Manufactured Product
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
NF - Total Alkali 95.0 - 100.5 %
NF - Content of Sodium 54.0 - 59.8 %
NF - Insoluble Substances and Organic Matter Passes Test
NF - Identification A (Sodium) Passes Test
NF - Identification B (pH) ≥11.0
NF - Potassium (K) ≤0.5 %
NF/FCC - Carbonate (as Na₂CO₃) ≤3.0 %
BP/EP - Assay (Total alkali as NaOH) 97.0 - 100.5 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Carbonate (as Na₂CO₃) ≤2.0 %
EP/BP - Chloride (Cl) ≤200 ppm
EP/BP - Sulfate (SO₄) ≤200 ppm
Appearance (white hygroscopic pellets)
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR