This medical-grade power strip supplies power to movable plug-connected components mounted to a rack, table or pedestal in operating rooms, patient-care vicinities, therapy rooms, imaging environments and administrative areas.
- Approved for use in patient-care vicinities when properly mounted
- Cord (10') with NEMA 5-15P-HG hospital-grade plug
- Features four NEMA 5-15R-HG hospital-grade outlets
- Optional drip shield provides extra protection from liquids
Two built-in resettable 15 A circuit breakers control power to all outlets and protect the power strip against overloads. Switchless design prevents the unit from being accidentally shut off. The built-in cord wrap provides easy cord management in mobile applications. The drip shield can be mounted for extra protection when there are liquids in the environment.
Certificaciones: Complies with UL 1363A specifications.
Información accesorios: HG outlet cover compatible with all NEMA 5-15R-HG and NEMA 5-15R outlets. Its self locking feature allows for easy installation and plugs are removable, reusable and designed for repeated use.
Información para pedidos: This product is covered by a lifetime limited warranty.