VWR® OR Towels
CT-02B
76321-390CS 179.61 USD
76321-390
VWR® OR Towels
OR Towels are highly absorbent and virtually lint-free.
  • Pre-washed, 100% cotton
  • Latex-frree
All sterile towels are CSR wrapped and have a special corner fold that allows for aseptic presentation into a sterile field.