Revco™ RDE Series –86 °C Ultra-Low Temperature Freezers with LN2 Back-up, Thermo Scientific
Supplier: Thermo Scientific
Revco®
RDE50086FDRLN RDE40086FARLN RDE60086FDRLN RDE40086FDRLN RDE50086FARLN RDE60086FARLN
76313-944EA 134541.98 USD
76313-944 76313-938 76313-948 76313-940 76313-942 76313-946
Revco™ RDE Series –86 °C Ultra-Low Temperature Freezers with LN2 Back-up, Thermo Scientific
Freezers Ultra Low Temperature Freezers
Dependable –86 °C storage, powered by H-drive for operational savings and sustainability.
  • Low energy
  • Low peak variation
  • Fast recovery after door opening
  • Long warm up time
  • User friendly and quiet
Designed to keep your samples safe. Limit environmental impact and lower HVAC costs.
Certificaciones: SNAP Compliant
Información de suministro: Package includes two shelves of sliding drawer racks and LN2 backup system.