Purging Manifold - The 'Werner', S.C.A.T.
Supplier: S.C.A.T.
106660
76316-230EA 486.4 USD
76316-230
Purging Manifold - The 'Werner', S.C.A.T.
Install up to four Safety Caps on your purging bottle with The 'Werner' - S.C.A.T. Europe's purging manifold.
  • Safe and easy handling
  • Capillaries are reliably fixed
  • Solvent filters have enough space due to wide GLS80 opening
Put your HPLC system into a clean initial state after the analytical process! The S.C.A.T. System for Solvent Safety stays permanently closed, keeping the 'safety chain' intact during the purging process.
No danger, no contamination, protect your lab personnel, the environment, and your HPLC system.