Sodium chloride 3% USP (Hypertonic)

Supplier: B BRAUN MEDICAL

Synonyms: HaliteSea saltSaline

L8051
76313-496CS 214.84 USD
76313-496
Sodium chloride 3% USP (Hypertonic)
Sodium chloride

3% Sodium Chloride Injection USP are sterile, nonpyrogenic and contain no bacteriostatic or antimicrobial agents. This intravenous solution is indicated for use in adults and pediatric patients as source of electrolytes and water for hydration.


  • NDC number: 00264-7805-10


Not manufactured with latex, PVC or DEHP


Specific Gravity: 1.021

Formula: NaCl
MW: 58.44 g/mol
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR