Dextrose 10% USP for injection
Supplier: B BRAUN MEDICAL
Synonyms:
Corn sugar
, D-Glucose
, D-(+)-Dextrose
, Glucolin
, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
, Glucose
, Dextrose
Dextrose Injections USP are sterile, nonpyrogenic and contain no bacteriostatic or antimicrobial agents. These products are intended for intravenous administration.
These intravenous solutions are indicated for use in adults and pediatric patients as sources of calories and water for hydration.
Not made with latex, PVC or DEHP.
Specific Gravity: 1.034
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol |
MDL Number:
MFCD00148912 CAS Number: 50-99-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...