HyClone™ Cosmic Calf Sera, New Zealand, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
SH30413.03IR25-40 SH30413.02IR SH30413.02HI SH30413.02
76236-370EA 177.67 USD
76236-370 82024-636 76236-372 76236-368
HyClone™ Cosmic Calf Sera, New Zealand, HyClone products (Cytiva)
HyClone™ Cosmic calf serum is fortified with iron and growth promoting factors.
  • Filtered through sequential 100 nm pore size rated filters
  • Extensive quality, performance and analytical testing