M9 Minimal Media Agar Plates, without Glucose, Teknova
101320-334
: Avantor
:
- Description:M9 Minimal Media Agar Plates, without Glucose
- Size:20 Plates
- Cat. No.:101320-334
- Plate diameter:100 mm
- Supplier no.:M1502
M9 minimal medium (M9) is commonly used for the cultivation of bacteria. It is comprised of a M9 minimal salts base formulation that can be supplemented with various amino acids and carbon sources.
M9 minimal medium (M9) contains 1X M9 salts, 0.2 mM MgSO4, 0.1 mM CaCl2, 1 mM thiamine and 1.5% agar.