Calibrated Glass Pipets, SUN SRi
Supplier: SUN SRi
205 044 205 024 205 584 205 592
14215-992CS 382.62 USD
14215-992 14234-450 14232-352 14232-356
Calibrated Glass Pipets, SUN SRi
Pipets Graduated Pipets
Calibrated glass pipets are manufactured from type 1, class A borosilicate glass.
  • All pipets have standard size tip openings
Certificaciones: Standard size pipets meet ASTM E 969 tolerance limits.