NitriDerm® EP Nitrile Examination Gloves with Extended Cuff, 182 Series, Innovative Healthcare
Supplier: INNOVATIVE HEALTHCARE CORP. TE
Certificates
About this item
Powder-free nitrile synthetic glove has an extended cuff length to provide extra protection in high risk situations.
- Excellent donning properties in a soft, low modulus formula
- Textured, slightly tacky surface for a better grip
- Thicker and longer than regular exam gloves
- Non-latex
- Textured finish
- No risk of latex allergens
- Improved wet/dry grip
Certifications: Complies with ASTM D6319, ASTM D5151, ASTM F1671.
Product Information
- Catalog #Supplier #Glove sizeGlove colorLengthThicknessAvailabilityPrice
- 76237-901182100SmallBlue11.5"5.5 mil.Box of 100$35.66Case of 1000$300.86Specifications:
Specifications
Environmentally Preferable - 76237-903182200MediumBlue11.5"5.5 mil.Box of 100$32.49Case of 1000$275.36Specifications:
Specifications
Environmentally Preferable - 76237-905182300LargeBlue11.5"5.5 mil.Box of 100$36.05Case of 1000$304.06Specifications:
Specifications
Environmentally Preferable - 76239-9451824002X-LargeBlue11.5"5.5 mil.Box of 100$33.70Case of 1000$304.06Specifications:
Specifications
Environmentally Preferable