J.T.Baker® BAKER INSTRA-ANALYZED®, AA Standards, Boron
Supplier: Avantor
J.T.Baker® BAKER INSTRA-ANALYZED®
BAKR6446-04 BAKR6446-04
JT6446-4CS 831.05 USD
CAJT6446-4 JT6446-4
J.T.Baker® BAKER INSTRA-ANALYZED®, AA Standards, Boron
Standards Cationic Standards Boron Standards
Boron standard solution for AAS are prepared from metals and salts of 99,999% spectral purity.
Prepared from H₃BO₃H
Certificaciones: Traceable to NIST and SRM numbers.