Sheep Gamma Globulin Fraction
Lyophilized Gamma Globulin Fraction
- prepared from normal serum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography
- control reagent
- Western Blotting and ELISA experiments
Sheep gamma globulin blocking reagent is an ideal blocker for western blotting, ELISA, Immunohistochemistry and other detection assays. Gamma globulins are a class of globulins, identified by their position after serum protein electrophoresis. The most significant gamma globulins are immunoglobulins ("Igs"), more commonly known as antibodies, although some Igs are not gamma globulins, and some gamma globulins are not Igs. Globulin is one of the three main types of serum proteins, the others being albumin and fibrinogen. Some globulins are produced in the liver, while others are made by the immune system.
Sheep Gamma Globulin Fraction can be utilized in Western Blotting and ELISA experiments.
- Catalog No:
- 75931-854
- 75931-852
- Antigen Symbol:
- Gamma Globulin
- Gamma Globulin
- Shipping Temperature:
- Ambient
- Ambient
- SizeSupplier no.
Specifications
Antigen SymbolGamma GlobulinEnvironmentally PreferableReactivityHostIsotypeShipping TemperatureAmbientSpecifications
Antigen SymbolGamma GlobulinEnvironmentally PreferableReactivityHostIsotypeShipping TemperatureAmbient