Recombinant Rat IL-6 control protein
- Control Protein
- PCA Assays
- ImmunoBlot
Interleukin 6 (IL-6) is an important pro-inflammatory and anti-inflammatory cytokine expressed by T cells, macrophages and muscle cells. IL-6 signals through a receptor complex containing two receptors, IL-6Rα and gp130. IL-6 has an important function in promoting fever and can serve to stimulate an immune response to trauma. IL-6 is often used for growth of hybridoma cell lines. Human IL-6 is active on mouse and rat cells. Recombinant rat IL-6 is a non-glycosylated protein, containing 188 amino acids, with a molecular weight of 21.8 kDa.
Interleukin-6 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-6 in immunological assays.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Source:
- Species:
- Environmentally Preferable:
- SizeSupplier no.
- 75929-338012-F01-310-001010 µg012F01310-0010Each (10µG)$352.09
- 75929-342012-F01-310-0100100 µg012F01310-0100Each (100µG)$1,160.75