United States
Order Entry
Anti-CD34 Rabbit Monoclonal Antibody [clone:013] APC (Allophycocyanin)
 
undefined
Anti-CD34 Rabbit Monoclonal Antibody [clone:013] APC (Allophycocyanin)
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Catalog No:
  • 103630-116
  • 103632-280
  • Antigen Symbol:
  • CD34
  • CD34
  • Antigen Name:
  • CD34 molecule
  • CD34 molecule
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • 013
  • 013
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Flow Cytometry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Storage Buffer:
  • PBS solution containing 0.5% BSA and 0.03% ProClin300
  • PBS solution containing 0.5% BSA and 0.03% ProClin300
  • Shipping Temperature:
  • Ambient
  • Ambient

 

 

  •  
     
    Size
     
     
  • 103630-116
    50589-R013-A-25
    25 tests
       
       
    Each
    Each - $90.90
    $90.90
     
    Each
       $90.90

     
     
     
    Specifications
    Antigen Symbol
    CD34
    Antigen Name
    CD34 molecule
    Environmentally Preferable
    Antibody Type
    Primary
    Clonality
    Monoclonal
    Conjugation
    APC (Allophycocyanin)
    Clone
    013
    Reactivity
    Mouse
    Host
    Rabbit
    Isotype
    IgG
    Flow Cytometry
    Yes
    Form
    Liquid
    Storage Buffer
    PBS solution containing 0.5% BSA and 0.03% ProClin300
    Shipping Temperature
    Ambient
  • 103632-280
    50589-R013-A-100
    100 tests
       
       
    Each
    Each - $282.80
    $282.80
     
    Each
       $282.80

     
     
     
    Specifications
    Antigen Symbol
    CD34
    Antigen Name
    CD34 molecule
    Environmentally Preferable
    Antibody Type
    Primary
    Clonality
    Monoclonal
    Conjugation
    APC (Allophycocyanin)
    Clone
    013
    Reactivity
    Mouse
    Host
    Rabbit
    Isotype
    IgG
    Flow Cytometry
    Yes
    Form
    Liquid
    Storage Buffer
    PBS solution containing 0.5% BSA and 0.03% ProClin300
    Shipping Temperature
    Ambient