Mouse Recombinant CSF1R/MCSF Receptor/CD115 Protein (His Tag) (from HEK293 Cells)
A DNA sequence encoding the mouse CSF1R (NP_001032948.2) extracellular domain (Met 1-Ser 511) was fused with a polyhistidine tag at the C-terminus.
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Immobilized M-CSF Protein, Mouse, Recombinant (ECD)(Cat:51112-MNAH) at 2 μg/ml (100 μl/well) can bind CSF1R Protein, Mouse, Recombinant (His Tag)(Cat:50059-M08H),the EC50 is 180-640 ng/ml.
- Gene ID:NP_001032948.2
- Reconstitution Instructions:A hardcopy of COA with reconstitution instruction is sent along with the products, please refer to it for detailed information
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:CD115|CSFR|HDLS|CSF-1R|C-FMS|MCSF Receptor|M-CSF-R|FMS|FIM2
- Protein/Peptide Name:CSF1R
- Purity:>97% as determined by SDS-PAGE
- Molecular Weight:The secreted recombinant mouse CSF1R consists of 503 amino acids and has a predicted molecular mass of 56.8 kDa. As a result of glycosylation, the apparent molecular mass of rm CSF1R is approximately 80-90 kDa in SDS-PAGE under reducing conditions.
- Sequence:Met1-Ser511
- Formulation:Lyophilized from sterile PBS, pH 7.4
- Shipping Temperature:Ambient
- Size