Epithelial Voltohmmeter, World Precision Instruments
Supplier: World Precision Instruments
EVOM2
MSPP-EVOM2EA 3772.31 USD
MSPP-EVOM2
Epithelial Voltohmmeter, World Precision Instruments
Electrical Meters Digital Multimeters
The EVOM is the instrument designed specifically to perform routine trans epithelial electrical resistance (TEER) measurement in tissue culture research.
  • Manual TEER measurements of epithelial cells in 6, 12, 24 and 96 well plates
  • Electrically isolated meter that plugs into a standard outlet for continual readout without push buttons
  • Compatible with endohm chambers
  • BNC output for data acquisition system
  • Auto ranging from 0 to 10 KΩ
  • Analog output for recording resistance or voltage measurements
  • Measures trans-epithelial electrical resistance or trans-epithelial voltage
  • Rechargeable battery allows up to 10 hours of mobile use
The instrument not only qualitatively measures cell monolayer health, but also quantitatively measures cellular confluence. The isolated power source is specifically designed to avoid adverse effects on tissue and the formation of electrode metal deposits, even when it is plugged into a standard wall outlet.
Power supply by internal rechargeable 6 V NiMH 2700 mAH battery with external 12 VDC supply for recharging.
Información de suministro: Includes industry standard STX2 hand held chopstick electrodes. 96-well plate measurement requires an STX100 series electrode, needs to be ordered seperately.