Nitrogen Generator, Genius NM32LA, Peak Scientific
Supplier: Peak Scientific
10-6020 10-6520
103549-798EA 19591.01 USD
103549-798 103549-796
Nitrogen Generator, Genius NM32LA, Peak Scientific
Gas Generators Nitrogen Generators
The culmination of over a decade's work perfecting on-site gas generation for LC-MS, the Genius NM32LA is the pinnacle of our Genius series.
  • Compressor based solution, no need for an external air supply
  • Service indication to allow planning of preventative maintenance
  • High-specification latest generation compressors located in insulated chamber reducing noise and vibration
  • Minimal set-up required
  • Highly economical source of nitrogen gas with low lifetime running costs
  • Proven robust and reliable nitrogen gas generation technology
  • Safe and practical laboratory-grade nitrogen
  • 12 month comprehensive on-site warranty
The NM32LA delivers up to 32 liters per minute of nitrogen gas, suitable for most LC-MS (liquid chromatography and mass spectrometry) instruments.
The culmination of over a decade's work perfecting on-site gas generation for LC-MS, the Genius NM32LA is the pinnacle of the Genius series. With thousands of units in the field, the NM32LA has become the proven and reliable nitrogen generator of choice in countless laboratories across the world.
The NM32LA delivers up to 32 liters per minute of nitrogen gas, suitable for most LC-MS instruments available on the market today.