VWR® Sterilized Direct Vent Soft Side Goggles
Supplier: VWR, part of Avantor
VWR®
515FFS
89522-676CS 883.66 USD
89522-676
VWR® Sterilized Direct Vent Soft Side Goggles
Goggles
Sterilized direct vent soft side goggles allow for a cool and comfortable fit.
  • Fog-free lens ensures a clear view
  • Blue tint with a blue rubber band strap
  • Packed and sealed in a Tyvek pouch
Featuring a fog-free lens, blue tint and blue rubber band head strap, these goggles allow users to stay protected while working.
Residuals (over eight-hour exposure period), Ethylene Chlorohydrin: 0.0429 mg
Residuals (over eight-hour exposure period), Ethylene Glycol: 0.1035 mg
Residuals (over eight-hour exposure period), Ethylene Oxide: <0.200 mg