Produced from sera of rabbits immunized with highly pure Recombinant Murine VEGF. AntiMurine VEGFspecific antibody was purified by affinity chromatography and then biotinylated.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Secondary
Antigen: VEGF
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 10776-280
- 10776-282
- Antigen Symbol:
- VEGF
- VEGF
- Antigen Name:
- VEGF
- VEGF
- Antibody Type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Murine VEGF
- Recombinant Murine VEGF
- Form:
- Lyophilized
- Lyophilized
- Molecular Weight:
- 39.0 kDa
- 39.0 kDa
- Purification:
- Recombinant Murine VEGF
- Recombinant Murine VEGF
- Storage Buffer:
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- Storage Temperature:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping Temperature:
- Ambient
- Ambient
- SizePackagingSupplier no.
- 10776-280500-P131BT-25UG25 µgPP Vial500P131BT25UGEach (25µG)$339.80
Specifications
Antigen SymbolVEGFAntigen NameVEGFEnvironmentally PreferableAntibody TypeSecondaryClonalityPolyclonalConjugationBiotinReactivityMouseHostRabbitIsotypeIgGWestern BlotYesELISAYesImmunogenRecombinant Murine VEGFFormLyophilizedMolecular Weight39.0 kDaPurificationRecombinant Murine VEGFStorage BufferSee Product Data Sheet for more information.Storage TemperatureStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient - 10776-282500-P131BT-50UG50 µgPP Vial500P131BT50UGEach (50µG)$434.63
Specifications
Antigen SymbolVEGFAntigen NameVEGFEnvironmentally PreferableAntibody TypeSecondaryClonalityPolyclonalConjugationBiotinReactivityMouseHostRabbitIsotypeIgGWestern BlotYesELISAYesImmunogenRecombinant Murine VEGFFormLyophilizedMolecular Weight39.0 kDaPurificationRecombinant Murine VEGFStorage BufferSee Product Data Sheet for more information.Storage TemperatureStore at –20 °C prior to reconstitution.Shipping TemperatureAmbient