United States
Order Entry
Anti-TRAIL Rabbit Polyclonal Antibody (Biotin)
 :  Peprotech
 
Anti-TRAIL Rabbit Polyclonal Antibody (Biotin)
 :  Peprotech
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Catalog No:
  • 10781-624
  • 10781-622
  • Antigen Symbol:
  • TRAIL
  • TRAIL
  • Antigen Name:
  • TRAIL
  • TRAIL
  • Antibody Type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Murine TRAIL
  • Recombinant Murine TRAIL
  • Form:
  • Lyophilized
  • Lyophilized
  • Molecular Weight:
  • 20.0 kDa
  • 20.0 kDa
  • Purification:
  • Recombinant Murine TRAIL
  • Recombinant Murine TRAIL
  • Storage Buffer:
  • See Product Data Sheet for more information.
  • See Product Data Sheet for more information.
  • Storage Temperature:
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Shipping Temperature:
  • Ambient
  • Ambient

 

 

  •  
     
    Size
    Packaging
    Supplier no.
     
     
  • 10781-624
    500-P303BT-50UG
    50 µg
    PP Vial
    500P303BT50UG
       
       
    Each (50µG)
    Each (50µG) - $434.63
    $434.63
    Each (50µG)
       $434.63

     
     
     
    Specifications
    Antigen Symbol
    TRAIL
    Antigen Name
    TRAIL
    Environmentally Preferable
    Antibody Type
    Secondary
    Clonality
    Polyclonal
    Conjugation
    Biotin
    Reactivity
    Mouse
    Host
    Rabbit
    Isotype
    IgG
    Western Blot
    Yes
    ELISA
    Yes
    Immunogen
    Recombinant Murine TRAIL
    Form
    Lyophilized
    Molecular Weight
    20.0 kDa
    Purification
    Recombinant Murine TRAIL
    Storage Buffer
    See Product Data Sheet for more information.
    Storage Temperature
    Store at –20 °C prior to reconstitution.
    Shipping Temperature
    Ambient
  • 10781-622
    500-P303BT-25UG
    25 µg
    PP Vial
    500P303BT25UG
       
       
    Each (25µG)
    Each (25µG) - $339.80
    $339.80
    Each (25µG)
       $339.80

     
     
     
    Specifications
    Antigen Symbol
    TRAIL
    Antigen Name
    TRAIL
    Environmentally Preferable
    Antibody Type
    Secondary
    Clonality
    Polyclonal
    Conjugation
    Biotin
    Reactivity
    Mouse
    Host
    Rabbit
    Isotype
    IgG
    Western Blot
    Yes
    ELISA
    Yes
    Immunogen
    Recombinant Murine TRAIL
    Form
    Lyophilized
    Molecular Weight
    20.0 kDa
    Purification
    Recombinant Murine TRAIL
    Storage Buffer
    See Product Data Sheet for more information.
    Storage Temperature
    Store at –20 °C prior to reconstitution.
    Shipping Temperature
    Ambient