LacSwitch II, Vectors, Agilent Technologies
Supplier: Agilent Technologies
217450 217450
95040-814EA 943.74 USD
95040-814 CA95040-814
LacSwitch II, Vectors, Agilent Technologies
Vectors, Plasmids and Libraries
The LacSwitch II inducible mammalian expression system utilizes an improved vector system in which several elements of the lac operon have been modified for use in eukaryotic cells for the control of gene expression.
pCMVLacI has CMV promoter for tighter repression of the inserted gene of interest.
pOPRSVI/MCS has 8 unique sites for directional insertion of gene of interest.
Información de suministro: Includes 100 µg pCMVLacl vector (1 mg/ml), 10 ug pOPRSVI/MCS vector (1 mg/ml), 10 ug pOPI3CAT vector (1 mg/ml), 1 g IPTG, Host strain: XL1-Blue MR
Precaución: For Research Use Only. Not for Use in Diagnostic Procedures.