Compatible with most plastic and glass pipettes from 0.1 to 100 ml.
- Eight speeds of selection for aspiration and dispensing different liquids
- Large LCD display shows speed settings and low battery warning
- Enables single-handed operation with minimum effort
- Lightweight and ergonomic design provides easy usability
- High-capacity Li-ion battery enables long runtime and extended operation
- Powerful pump fills a 25 ml pipette in less than 5 seconds
- 0.45 μm replaceable hydrophobic filter
- Rechargeable during use
A 2 to 3 hour charge time enables up to 8 hours of continuous use.
Certificaciones: CE and cTUVus.
Información de suministro: The Levo Plus® pipette filler is supplied with battery,1-piece stand, 1-piece charger, and a power adaptor.