LigaTrap® Sheep IgG Purification Kit, ImmunoReagents
LT-141 LT-141KIT
75783-520EA 654.23 USD
75783-520 75783-532
LigaTrap® Sheep IgG Purification Kit, ImmunoReagents
LigaTrap® Sheep IgG Purification Kit uses LT technology to utilize novel, patented ligands, designed for antibody purification.
LigaTrap® Sheep IgG Purification Resin binds an exceptionally broad range of recombinant, monoclonal and human or animal source polyclonal immunoglobulins.
Información de suministro: Kit includes ten spin columns filled with LigaTrap® resin and buffers needed to capture species-specific antibodies.