LigaTrap® Llama IgG Purification Kit, ImmunoReagents
LT-144KIT LT-144
76199-394EA 379.04 USD
76199-394 76199-396
LigaTrap® Llama IgG Purification Kit, ImmunoReagents
LigaTrap® Llama IgG Purification Kit uses LT technology to utilizes novel, patented ligands, designed for antibody purification.
LigaTrap® Llama IgG Purification Resin binds an exceptionally broad range of recombinant, monoclonal and human or animal source polyclonal immunoglobulins.
Información de suministro: Kit includes ten spin columns filled with LigaTrap® resin and buffers needed to capture species-specific antibodies.