These microscopes are ideal for biological sciences, dissections, geology, paleontology, food science, small device assembly or gross observation in small detail.
- Durable, cost effective microscope to provide crisp and clear images
- Deliver brilliant contrast, parfocality, flatness and depth of field
- Auxiliary lenses and eye pieces can increase or decrease both magnification and working distance/field of view
- No need to refocus when zooming magnification, the GL Series is parfocal
These microscopes minimize fatigue with flat field of view, side mounted zoom controls, low positioned focus control, 45° ocular incline all work together to reduce stress and allow work in a natural position.
Accommodate all users with wide interpupillary adjustment (45 to 75 mm) and diopters in both high eye point eyepieces allow virtually all individuals comfort in viewing.
Información de suministro: Includes a pair of 10x eye pieces, black/white reversible, stage plate, dust cover, power cord and instruction manual.