Would you like to log in again?
Vitality hrGFP Mammalian Expression, Agilent Technologies
Supplier: AGILENT TECH
240142 240242 240241 240036 240032 240142 240062 240141 240042 240062 240241 240059 240063 240032 240242 240035 240141 240043 240035 240044 240043 240036 240042 240044
99901-068EA 749.7 USD InStock
CA97066-374 99901-068 99901-060 97066-340 CA97066-336 97066-374 CA97066-358 CA97066-372 97066-342 97066-358 CA99901-060 97066-356 97066-360 97066-336 CA99901-068 97066-338 97066-372 97066-344 CA97066-338 CA97066-346 CA97066-344 CA97066-340 CA97066-342 97066-346
Vitality hrGFP Mammalian Expression, Agilent Technologies
Protein Expression Systems
Vitality hrGFP (humanized, Renilla reniformis green fluorescent protein) products are optimal for mammalian expression protein studies that examine gene expression, transfection efficiency, and protein localization.
  • Applications include Western blotting, immunoprecipitation and flow cytometry
This hrGFP is derived from a different organism than EGFP, so the hrGFP antibody does not cross react with the Aequorea victoria GFP variant-derived EGFP.
Precaución: For Research Use Only. Not for Use in Diagnostic Procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty