Puerto Rico
Order Entry
Recombinant Human Proprotein Convertase 9, Bon Opus Biosciences
Recombinant Human Proprotein Convertase 9, Bon Opus Biosciences
 
Recombinant Human Proprotein Convertase 9, Bon Opus Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
  • Protein/Peptide Name:
    Proprotein Convertase Subtilisin/Kexin Type 9

 

 

  •  
     
    Size
    Supplier no.
     
     
  • 102943-632
    CA85-1MG
    1 mg
    CA85-1MG
       
       
    Each (1mg)
    Each (1mg)

     
  • 102943-630
    CA85-500UG
    500 μg
    CA85-500UG
       
       
    Each (500µG)
    Each (500µG)

     
  • 102943-628
    CA85-50UG
    50 μg
    CA85-50UG
       
       
    Each (50µG)
    Each (50µG)

     
  • 102943-626
    CA85-10UG
    10 μg
    CA85-10UG
       
       
    Each (10µG)
    Each (10µG)