SP Wilmad-LabGlass O-Rings, PTFE, SP Industries
Supplier: SP Industries
LG-1022-471 LG-1022-402 LG-1022-416 LG-1022-458 LG-1022-414 LG-1022-420 LG-1022-434 LG-1022-406 LG-1022-422 LG-1022-478 LG-1022-432 LG-1022-412 LG-1022-446 LG-1022-454 LG-1022-400 LG-1022-404 LG-1022-428 LG-1022-438 LG-1022-462 LG-1022-452 LG-1022-450 LG-1022-426
14211-650EA 2.84 USD
14211-650 14201-414 14211-564 14209-718 14201-388 14211-636 14211-668 14211-568 14201-584 14201-376 14211-644 14212-090 14212-082 14201-374 14211-616 14212-084 14211-648 14212-048 14212-058 14212-038 14206-980 14210-746
SP Wilmad-LabGlass O-Rings, PTFE, SP Industries
These polytetrafluoroethylene (PTFE) O-Rings come in a variety of sizes. PTFE offers excellent chemical resistance.
  • With a density of 2.20 g/cm³, it is opaque white, and has the lowest coefficient of friction of any solid
  • Useful temperature range is –180 to 260 °C
  • Virtually inert, but not recommended for use with fluorine, gaseous, or molten alkali metals