PhosphoSeek™ Phosphoprotein Enrichment Kit, BioVision, Inc.
Supplier: BioVision
K1402-6 K1402-2
76211-784EA 445.01 USD
76211-784 76211-786
PhosphoSeek™ Phosphoprotein Enrichment Kit, BioVision, Inc.
Protein Purification Protein Purification Kits
IMAC based purification of phosphorylated proteins
  • Simple procedure and can purify phosphoprotein within 1 hour.
  • The kit provides unique formulations of buffers and column to enrich phosphorylated proteins upto 5-10 folds
  • The purified sample can directly be analyzed by two-dimensional electrophoresis or mass spectrometry.
PhosphoSeek™ Phosphoprotein Enrichment Kit is based on IMAC (immobilized metal affinity chromatography), can isolate the most amount of phosphoprotein from different biological materials, such as animal, plant and microorganism. Isolated phosphoprotein can be Ideal for use in many downstream applications, including mass spectrometry and 2D-PAGE.