Order Entry
Econo™ Lahey Gall Duct Forceps, Floor Grade, Sklar®
  76192-482
 :  Sklar
undefined
Econo™ Lahey Gall Duct Forceps, Floor Grade, Sklar®
Catalog # 76192-482
Supplier:  Sklar
Supplier Number:  21-453
Price
$190.35
Log in for availability
Unit of Measure Box of 12
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Length:
    7.5"
  • Material:
    Stainless steel
  • Curvature:
    Curved
  • Cat. No.:
    76192-482
Econo™ Lahey Gall Duct Forceps are used for blunt dissection and for clamping vessels.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Econo™ Lahey Gall Duct Forceps have longitudinal serrations on curved tips. The curvature of this instrument allows for dissection in areas that may not be easily accessible. They have ratcheted finger rings for locking into place during procedures. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.