Vacuum Sealer, 35 Series, SCS
35VACUSEAL20 35VACUSEAL30
89024-424EA 8285.16 USD
89024-424 89024-428
Vacuum Sealer, 35 Series, SCS
A vacuum sealer removes moisture laden air from the interior of moisture barrier bags, and heat seals the bags closed.
  • A fixed position nozzle evacuates the bag
  • No vacuum pump
Timers for vacuum, sealing, and cooling are activated by a single foot pedal, and insure that the bag is evacuated and sealed.
Información para pedidos: Vacuum sealers cannot be returned unless a manufacturing defect is verified by 3M. An RMA number must be obtained prior to any return. Gas purge option and support tray are available.