Kimberly-Clark™ Isolation Cover Gown, Kimberly-Clark Professional
Supplier: Kimberly-Clark
69981
76204-808CS 431.48 USD
76204-808
Kimberly-Clark™ Isolation Cover Gown, Kimberly-Clark Professional
Kimberly-Clark™ isolation cover gowns are made for facility-wide use when risk of exposure to fluid is between low and moderate.
  • Protective 3-layer SMS fabric with light to moderate fluid protection
  • Tape-tab neck closure with gathered back
  • Tie waist and elastic cuff
  • Gathered back
Isolation cover gowns are made of a medium-weight SMS fabric and are for facility-wide use when risk of exposure to fluid is between low and moderate. Recommended areas of use include ICU, hyperbaric and dialysis units, contact isolation, med/surgery units, nursery, labs, and pathology facilities.
Certificaciones: Meets AAMI Level 2 requirements.