Labeling 5X Buffer, Promega
Supplier: Promega
U1151
PAU1151EA 48.3 USD
PAU1151
Labeling 5X Buffer, Promega
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
A complete set of complementary reagents, including Labeling 5X Buffer that contains random synthetic hexadeoxynucleotide primers for random-primed labeling of linear template DNA with radionucleotides.
  • Complete Kit for Random-Primed Labeling of Linear Template DNA, Excluding Radionucleotides
  • Ready-to-use reagents for random-primed labeling of linear DNA, including random hexamer primers (excludes radionucleotides)
  • Probes generated with high specific activities >1 x 109cpm/microg
As little as 25ng of input DNA can be used to generate probes with specific activities >1 x 10(9)cpm/ug.
Precaución: Storage Temperature -30°C to -10°C