Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lico Spectral Colorimeters, Hach

Supplier: Hach
LMV1875320001 LMV1875340001
Competence in Color.

  • All important color scales included in one instrument
  • Simple integration into the laboratory network through Ethernet connection
  • High level of measurement reliability through a comprehensive set of test aids
  • Easy handling

The spectral color measurement of clear liquids is indispensable in many fields and serves as a quality assurance method for solvents, paints and pharmaceuticals. The highest measuring reliability is essential. Hach® ensures simple, fast measurements for the user. More than 25 years of expertise in the developmentof colorimeters have gone into the Lico line of instruments.

All important color scales included in one instrument. The Lico 620 with up to 5 color scales is designed for fast routine measurements in the laboratory and in production facilities. It is already in use in a wide variety of areas in the chemical industries for quality and production control, e.g. to assess surfactants, oils, fats, resins and synthetic resins. The Lico 690 Spectral Colorimeter is flexible to use and works with many applications, such as chemical, pharmaceuticals, oil industries and the food and beverage industry. 26 color scales are directly integrated in the instrument: conventional scales like iodine, Hazen (Pt Co), Gardner, harmacopoeia as well as specific scales like Saybolt or ASTM.Simple integration into the laboratory network through Ethernet connection. Lico instruments can be easily integrated to existing laboratory networks through an Ethernet interface.

High level of measurement reliability through a comprehensive set of test aids Certified test filters, reproducible standard color solutions and an integrated implementation of AQA (Analytical Quality Assurance) ensure optimum measurement reliability. Measured values can be evaluated in all scales, even post-measurement with archived spectral data.

Easy handling–The measurement process starts automatically when the sample cell is inserted, and the measurement vial is identified by the instrument. The easy-to-change cell compartment facilitates cleaning and/or replacement. The operation of the Lico colorimeters requires only minimum training effort thanks to clear instructions on the color touchscreen.

Lico 620 color scales include DIN 6162 iodine; ISO 6271 Hazen (Pt/Co, APHA, ASTM D1209, D5386); ISO 4630 Gardner (ASTM D1544, D6045); ASTM D 156 Saybolt; ASTM D 1500 mineral oil.

Lico 690 color scales include European, US and Chinese Pharmacopoeia (EP, USP, CP); DIN 6162 Iodine; ISO 6271 Hazen (Pt/Co, APHA, ASTM D1209, D5386); ISO 4630 Gardner (ASTM D1544, D6045, D6166); ISO 27608 Animal and Vegetable Fats and Oils; ISO 11664 Colorimetry; ASTM E308 CIE Color; ASTM D156 Saybolt; ASTM D1500 Mineral Oil (ISO 2049); ASTM D848 Acid Wash Test; ASTM D1925 Yellowness Index (ASTM D5386); AOCS Cc13e, BS 684 Ly/Lr; ADMI, ICUMSA, EBC, ASBC, Hess-Ives.

Información para pedidos: Includes instrument, power supply, printed basic user manual, CD with full user manual in 4 languages, 10 mm cell adapter, dust cover. Accessories not included.



 

ORDER

Expand All / Collapse All
Description Memory Supplier No. VWR Catalog Number Unit Price Quantity
Lico 620 Colorimeter for up to Five Color Scales 400 Color Readings LMV1875320001 76200-298 Each Retrieving Restricted
Lico 690 Professional Spectral Colorimeter with Spectral Scan (320 – 1100 nm) and Sipper (Optional), 25+ Color Scales Available 100 Color References; 1000 Photometric Values;
20 Wavelength Scans; 20 Time Scans; 3000 Color
Readings
LMV1875340001 76200-300 Each Retrieving Restricted
restricted This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Optical design 0°/180° rectilinear
Source Lamp Tungsten halogen lamp
Wavelength Range 380 – 720 nm used for colorimetric calculations
Display TFT WVGA Color Graphic Touch Screen
Electrical 100 – 240 VAC
Display Dimensions 17.8 cm (7")
Beam Height 10 mm
Compatibility 10 mm Rectangular Cuvette;
11 mm Round Cuvette;
50 mm Rectangular Cuvette

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
Addista Color Standards; 3 x 50 mL Standard Liquid for Hazen Color (20,100,200) and Standard Liquid for Gardner Color (2,5,8) With Certificate for Lico, 12 Month Shelf Life LZM282 76200-302 Each Retrieving Restricted
Thermal Printer Paper for Citizien Pd-24; 4 Rolls of Printer Paper, 11 cm Width LZM078 76200-304 Pack of 4 Retrieving Restricted
Set of Test Filter; Set of Four Certified Test Glass Filters for Stray Light, Absorbance And Wavelength Test, 24 Month Shelf Life LZM339 76200-306 Pack of 4 Retrieving Restricted
Replacement Cell Compartment, 50 mm Rectangular LZM368 76200-310 Each Retrieving Restricted
Adapter for Cuvettes LZM369 76200-312 Each Retrieving Restricted
SIP 10 Sipper Set; Sipper Set for Flow Through Applications in UV Range. 1 cm Flow Through Cell, Quartz LQV1579930001 76200-314 Each Retrieving Restricted
Disposable Plastic Cuvettes, 10 x 10 mm, 1000 Pieces LYY214 76200-318 Pack of 1,000 Retrieving Restricted
Sample Cell, Rectangular, Glass, 10 mm, 190 – 1100 nm Wavelength Range, Pack of Three LZP045 76200-320 Pack of 3 Retrieving Restricted
Replacement Halogen Bulb LZV565 76200-322 Each Retrieving Restricted
Disposable Plastic Rectangular Cuvettes, 50 x 10 mm, 50 Pieces LZM130 76200-324 Pack of 50 Retrieving Restricted
Disposable Round Cuvettes, Glass, 11 mm, 560 Pieces LYY621 76200-326 Pack of 560 Retrieving Restricted
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty