Puerto Rico
Order Entry
Anti-MSN Mouse Monoclonal Antibody (CF647) [clone: MSN493]
Anti-MSN Mouse Monoclonal Antibody (CF647) [clone: MSN493]
 :  Biotium
 
Anti-MSN Mouse Monoclonal Antibody (CF647) [clone: MSN493]
 :  Biotium
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Catalog No:
  • 75885-928
  • 75885-926
  • Antigen Symbol:
  • MSN
  • MSN
  • Antigen Name:
  • Moesin
  • Moesin
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • MSN493
  • MSN493
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • Unigene ID: 87752
  • Unigene ID: 87752
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western Blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0.1 mg/mL
  • 0.1 mg/mL
  • Storage Buffer:
  • PBS/0.1% BSA/0.05% azide
  • PBS/0.1% BSA/0.05% azide

 

 

  •  
     
    Size
    Supplier no.
     
     
  • 75885-928
    BNC470493-500
    500 μL
    BNC470493-500
       
       
    Each (500µl)
    Each (500µl) - $569.73
    $569.73
    Each (500µl)
       $569.73

     
     
     
    Specifications
    Antigen Symbol
    MSN
    Antigen Name
    Moesin
    Environmentally Preferable
    Antibody Type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 647
    Clone
    MSN493
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    Unigene ID: 87752
    Isotype
    IgG1 kappa
    Western Blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoPrecipitation
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0.1 mg/mL
    Storage Buffer
    PBS/0.1% BSA/0.05% azide
  • 75885-926
    BNC470493-100
    100 μL
    BNC470493-100
       
       
    Each (100µl)
    Each (100µl) - $205.77
    $205.77
    Each (100µl)
       $205.77

     
     
     
    Specifications
    Antigen Symbol
    MSN
    Antigen Name
    Moesin
    Environmentally Preferable
    Antibody Type
    Primary
    Clonality
    Monoclonal
    Conjugation
    CF® 647
    Clone
    MSN493
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    Unigene ID: 87752
    Isotype
    IgG1 kappa
    Western Blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoPrecipitation
    Yes
    ImmunoFluorescence
    Yes
    Concentration
    0.1 mg/mL
    Storage Buffer
    PBS/0.1% BSA/0.05% azide