Specifications
- Description:GloMax-Multi Luminescence Module, 1 each
- Size:1 each
- Cat. No.:PAE7041
- Storage temperature:15...30 °C
- Supplier no.:E7041
Specifications
About this item
The GloMax-Multi Detection System consists of a base unit plus modular detection and functional units. While maintaining state-of-the-art luminescence performance, the GloMax line now extends into fluorescence and absorbance applications.
- Modules for the GloMax(R)-Multi Microplate Multimode Reader
- Instrument is discontinued
- Modules available until June 1, 2018
The GloMax-Multi Microplate Multimode Reader has been a workhorse instrument for the past several years, but as of June 1, 2013, we are retiring this instrument model. It continues to offer state-of-the-art luminescence multimode performance for customers worldwide. All accessories and modules will continue to be available, as will instrument service, until June 1, 2018. We kindly invite you to review your current and future laboratory needs relative to your GloMax-Multi Instrument. If you have any questions or would like an assessment based on your laboratory and budgetary needs, contact your local Promega Sales Representative today or visit us at: www.promega.com/detection-home to learn more about our new products, trade-in programs and service agreement options. The GloMax-Multi Detection System consists of a base unit plus modular detection and functional units, allowing for a flexible solution that can be expanded over time. Luminescence, fluorescence and absorbance reading modules are available as well as an optional injector system (used with the luminescence detection module only). Luminescence Module: An advanced photon-counting photomultiplier tube (PMT) provides unmatched signal-to-noise ratios, beating most standalone luminometers. The luminescence module can detect as little as 3 x 10(-21) moles of luciferase, covering a dynamic range over 8 logs. A dual masking system minimizes well-to-well cross-talk. Fluorescence Module: Application-optimized Optical Kits simplify fluorescence operation while maximizing performance. Long-lived LED-based excitation lights minimize maintenance and variability in intensity. The UV, Blue, Green and Red Optical Kits are included with the Fluorescence Detection Module. An AFC Optical Kit is available as an accessory. Absorbance Module: A 6-position filter wheel with 2 open positions ensures flexibility for a wide range of applications. An LED-based visible spectrum light source minimizes maintenance and variability. Filters for reading 450, 560, 600 and 750nm are included. A 490nm filter is available as an accessory. Operation of the GloMax-Multi Detection System can be performed entirely through the touch screen. Protocols for a variety of Promega and common laboratory assays are pre-installed, and there is also the flexibility to modify the protocols or create new methods. Data can be saved on the instrument and moved via the included USB flash drive. The system works as a standalone workstation in the laboratory, freeing computing resources from data capture, so more resources can be directed toward analysis. To learn more about the GloMax Systems, please visit: www.promega.com/products/instruments/